missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cystatin SA Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99787-100ul
This item is not returnable.
View return policy
Description
Cystatin SA Polyclonal antibody specifically detects Cystatin SA in Human samples. It is validated for ELISA
Specifications
| Cystatin SA | |
| Polyclonal | |
| ELISA 1:5000-1:10000 | |
| cystatin 2, cystatin S5, cystatin SA, Cystatin-2, Cystatin-S5, cystatin-SA, cysteine-proteinase inhibitor, MGC71924, salivary cysteine (thiol) protease inhibitor | |
| Produced in rabbits immunized with purified, recombinant Human Cystatin SA (Accession#: NP_001313.1; Met 1-Ala 141) | |
| 100 μg | |
| Cell Biology | |
| 1470 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction