missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cystatin SN Antibody (MM0234-5N51), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12255MFV450
This item is not returnable.
View return policy
Description
Cystatin SN Monoclonal antibody specifically detects Cystatin SN in Human samples. It is validated for Western Blot
Specifications
| Cystatin SN | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| MM0234-5N51 | |
| Western Blot | |
| cystain-SA-I, cystatin 1, cystatin SA-I, cystatin SN, cystatin-1, cystatin-SN, Salivary cystatin-SA-1, type 2 family | |
| Human recombinant Cystatin SN | |
| 0.1 mL | |
| Proteases & Other Enzymes | |
| 1469 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction