missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cystatin/Stefin Homolog Antibody (RM0218-8D33), Allophycocyanin, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-12412APC
This item is not returnable.
View return policy
Description
Cystatin/Stefin Homolog Monoclonal antibody specifically detects Cystatin/Stefin Homolog in Mouse samples. It is validated for Western Blot, Neutralization
Specifications
| Cystatin/Stefin Homolog | |
| Monoclonal | |
| APC | |
| PBS | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| 1475 | |
| Store at 4C in the dark. | |
| IgG2 |
| Western Blot, Neutralization | |
| RM0218-8D33 | |
| Western Blot, Block/Neutralize | |
| AREI, PSS4, STF1, STFA | |
| Mouse recombinant Cystatin/Stefin Homolog | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction