missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYTH2, Mouse anti-Human, Clone: 5E11, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00009266-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: VDDPRKPNCFELYIPNNKGQLIKACKTEADGRVVEGNHMVYRISAPTQEEKDEWIKSIQAAVSVDPFYEMLAARKKRISVKKKQESpecifications
| CYTH2 | |
| Monoclonal | |
| Unconjugated | |
| cytohesin 2 | |
| ARNO/CTS18/CTS18.1/PSCD2/PSCD2L/SEC7L/Sec7p-L/Sec7p-like | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 5E11 | |
| In 1x PBS, pH 7.4 | |
| NM_017457 | |
| CYTH2 | |
| CYTH2 (NP_059431, 314 a.a. ∼ 398 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 9266 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction