missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cytidine-5'-monophosphate disodium salt, 99+%

Catalog Number p-7042686
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
6757-06-8
C9H12N3Na2O8P
367.16
MFCD00150830
INTPYBRGLGSMRA-IJRYJGAHNA-L
cytidine 5'-monophosphate disodium salt, disodium cmp, cmp disodium salt, cytidine-5'-monophosphate disodium salt, disodium 5'-cmp, 5'-cmp disodium salt, disodium 5'-o-phosphonatocytidine, 5'-cytidylic acid, disodium salt, disodium cytidine 5'-monophosphate, cytidine monophosphate disodium salt
94332
disodium;[(2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate
[Na+].[Na+].NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]1O
This item is not returnable. View return policy
6757-06-8
C9H12N3Na2O8P
367.16
MFCD00150830
INTPYBRGLGSMRA-IJRYJGAHNA-L
cytidine 5'-monophosphate disodium salt, disodium cmp, cmp disodium salt, cytidine-5'-monophosphate disodium salt, disodium 5'-cmp, 5'-cmp disodium salt, disodium 5'-o-phosphonatocytidine, 5'-cytidylic acid, disodium salt, disodium cytidine 5'-monophosphate, cytidine monophosphate disodium salt
94332
disodium;[(2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate
[Na+].[Na+].NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]1O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Quantity 1 g
Formula Weight 367.16
Percent Purity ≥99%
Physical Form Crystalline Powder
Chemical Name or Material Cytidine-5'-monophosphate disodium salt

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'