Learn More
Invitrogen™ CYTIP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114192
Description
Immunogen sequence: QLALTRSSSL SDFSWSQRKL VTVEKQDNET FGFEIQSYRP QNQNACSSEM For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Guinea Pig: 75%; Human: 100%; Mouse: 79%; Pig: 79%; Rat: 79%.
The protein encoded by this gene contains 2 leucine zipper domains and a putative C-terminal nuclear targeting signal, but does not have any hydrophobic regions. This protein is expressed weakly in resting NK and T cells. The encoded protein modulates the activation of ARF genes by CYTH1. This protein interacts with CYTH1 and SNX27 proteins and may act to sequester CYTH1 protein in the cytoplasm.
Specifications
| CYTIP | |
| Polyclonal | |
| Unconjugated | |
| CYTIP | |
| A130053M09Rik; AI462064; B3-1; C80816; CASP; Cbp; cbp HE; CYBR; CYTHIP; Cytip; cytohesin 1 interacting protein; cytohesin binder and regulator; cytohesin binding protein; cytohesin binding protein HE; cytohesin-1 interacting protein; cytohesin-associated scaffolding protein; Cytohesin-binding protein HE; cytohesin-interacting protein; cytohesin-interacting protein-like; HE; Pleckstrin homology Sec7 and coiled-coil domain-binding protein; pleckstrin homology Sec7 and coiled-coil domains-binding protein; pleckstrin homology, Sec7 and coiled/coil domains, binding protein; pleckstrin homology, Sec7 and coiled-coil domains, binding protein; Pscdbp; Sec7; tamalin; zmp:0000001116 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 9595 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O60759 | |
| CYTIP | |
| Immunogen sequence: QLALTRSSSL SDFSWSQRKL VTVEKQDNET FGFEIQSYRP QNQNACSSEM (aa 58-107). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.