missing translation for 'onlineSavingsMsg'
Learn More

Cytochrome c Antibody (CYCS/1010), HRP, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-47693H

Product Code. 18667355

  • € 571.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Cytochrome c Monoclonal antibody specifically detects Cytochrome c in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cytochrome c
Monoclonal
HRP
CYCHCS, cytochrome c, cytochrome c, somatic, THC4
Recombinant full-length human Cytochrome c protein (Uniprot: P99999)
0.1 mL
Apoptosis, Cellular Markers, Cholesterol Metabolism, Core ESC Like Genes, Lipid and Metabolism, Mitochondrial Markers, Stem Cell Markers
54205
Store at 4°C in the dark.
IgG2b κ
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF
CYCS/1010
PBS
Mouse
Protein A or G purified
RUO
Primary
Human, Rat
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.