missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cytochrome C Chimeric Recombinant Rabbit Monoclonal Antibody (3.7C4)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA548254
This item is not returnable.
View return policy
Description
Specificity: This antibody recognises mouse cytochrome c (CYT) which is a part of the respiratory chain responsible for the catalysis of the reduction of oxygen to water.
Cytochrome C is an electron transporting protein that resides within the intermembrane space of the mitochondria, where it plays a critical role in oxidative phosphorylation and the production of cellular ATP. Release of cytochrome C from the mitochondria to the cytosol is triggered by apoptotic stimuli. In the cytoplasm, the cytochrome C binds apoptotic protease activating factor which activates the apoptotic initiator procaspase 9. Cytosolic cytochrome C functions in the activation of caspase 3, an ICE family molecule that is a key effector of apoptosis.
Specifications
| Cytochrome C Chimeric | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| P62897 | |
| Cycs | |
| This antibody was raised by immunising OVA-primed mice with cytochrome c-OVA. | |
| 200 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG κ |
| ELISA, Immunocytochemistry | |
| 3.7C4 | |
| Unconjugated | |
| Cycs | |
| CG17903; CG17903-PA; CG17903-PB; CRF; CYC; CYC2; Cycs; CYCSA; Cyct-c-p; Cyt c; cyt c-p; cyt.c; CytC; Cyt-c; CYTC2; Cyt-c2; CytC-2; cyt-c-p; cytC-p; Cyt-c-p-PA; Cyt-c-p-PB; Cytochrome c; cytochrome c DC4; Cytochrome c proximal; Cytochrome C, expressed in somatic tissues; cytochrome c, somatic; Cytochrome c-2; cytochrome c-like; cytochrome c-like protein; Cytochrome c-proximal; cytochrome-c-p; Cytochrome-c-proximal; DC4; dCyt.C-p; DMc01; Dmel\CG17903; Dmel_CG17903; hc1; HCS; l(2)k13905; lethal (2) k13905; LOC100341175; LOC100357840; MGC128356 protein; pDMc01; RH17228p; THC4 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 13063 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction