missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome P450 26B1 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-53810
This item is not returnable.
View return policy
Description
Cytochrome P450 26B1 Polyclonal specifically detects Cytochrome P450 26B1 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications
| Cytochrome P450 26B1 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| CYP26A2, Cytochrome P450 26A2, cytochrome P450 26B1, cytochrome P450 CYP26B1 variant 2, Cytochrome P450 retinoic acid-inactivating 2, cytochrome P450 retinoid metabolizing protein, cytochrome P450, family 26, subfamily B, polypeptide 1, Cytochrome P450RAI-2, DKFZp686G0638, EC 1.14, MGC129613, P450RAI2, P450RAI-2, Retinoic acid-metabolizing cytochrome | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Cross reacts with Human. Expected from sequence similarity: Mouse, Rat, Dog, Pig and Cow. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
| 0.5 mg/mL | |
| Flow Cytometry 10 μg/mL, Immunohistochemistry 4 - 6 μg/mL, Immunohistochemistry-Paraffin 4 - 6 μg/mL, Peptide ELISA Detection limit 1:128000 | |
| NP_063938.1 | |
| CYP26B1 | |
| Peptide with sequence C-SQARSEDKDGRFH corresponding to internal region according to NP_063938.1. | |
| 0.1 mg | |
| Cancer, Cardiovascular Biology, Cellular Signaling, metabolism | |
| 56603 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction