missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome P450 26B1 Antibody (T5P3G2*A3), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-50195CL1
This item is not returnable.
View return policy
Description
Cytochrome P450 26B1 Monoclonal antibody specifically detects Cytochrome P450 26B1 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| Cytochrome P450 26B1 | |
| Monoclonal | |
| CoraFluor 1 | |
| CYP26A2, Cytochrome P450 26A2, cytochrome P450 26B1, cytochrome P450 CYP26B1 variant 2, Cytochrome P450 retinoic acid-inactivating 2, cytochrome P450 retinoid metabolizing protein, cytochrome P450, family 26, subfamily B, polypeptide 1, Cytochrome P450RAI-2, DKFZp686G0638, EC 1.14, MGC129613, P450RAI2, P450RAI-2, Retinoic acid-metabolizing cytochrome | |
| Ovalbumin-conjugated synthetic peptide DSNQNEILPE (C-terminal sequence) | |
| 0.1 mL | |
| Cancer, Cardiovascular Biology, Cellular Signaling, metabolism | |
| 56603 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| T5P3G2*A3 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction