missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome P450 2A6, 2A7, 2A13 Antibody (F16 P2 D8), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50184FR
This item is not returnable.
View return policy
Description
Cytochrome P450 2A6, 2A7, 2A13 Monoclonal specifically detects Cytochrome P450 2A6, 2A7, 2A13 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| Cytochrome P450 2A6, 2A7, 2A13 | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ovalbumin-conjugated synthetic peptide RNYTMSFLPR. CYP2A6, CYP2A7and CYP2A13 have the same c-terminal sequence. (C-terminal sequence identical in human and rhesus macaque) | |
| 0.1 mL | |
| Cancer, Cardiovascular Biology, Cellular Signaling, metabolism | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry | |
| F16 P2 D8 | |
| Western Blot, ELISA, Immunohistochemistry | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction