missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome P450 2A6 Antibody (OTI1D2), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70524AF350
This item is not returnable.
View return policy
Description
Cytochrome P450 2A6 Monoclonal specifically detects Cytochrome P450 2A6 in Human, Mouse, Rat, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
| Cytochrome P450 2A6 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| CYP2A6 | |
| Full length recombinant protein of human Cytochrome P450 2A6 (NP_000753) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Primate, Monkey | |
| IgG2b |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| OTI1D2 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| Coumarin 7-hydroxylase, CPA6, CYP2A3P450C2A, CYP2AEC 1.14.14.1, CYPIIA6, cytochrome P450 2A6, Cytochrome P450 IIA3, Cytochrome P450(I), cytochrome P450, family 2, subfamily A, polypeptide 6, cytochrome P450, subfamily IIA (phenobarbital-inducible), polypeptide 6, flavoprotein-linked monooxygenase, P450PB, xenobiotic monooxygenase | |
| Mouse | |
| Affinity Purified | |
| Lipid and Metabolism | |
| 1548.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction