missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome P450 2B1, 2B2 Antibody (b/e3), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50201MFV450
This item is not returnable.
View return policy
Description
Cytochrome P450 2B1, 2B2 Monoclonal antibody specifically detects Cytochrome P450 2B1, 2B2 in Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications
| Cytochrome P450 2B1, 2B2 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rat liver cytochromes p450 2B1 and 2B2 | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, ELISA, Immunoprecipitation | |
| b/e3 | |
| Western Blot, ELISA, Immunoprecipitation | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction