missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome P450 2C18 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92808-0.02ml
This item is not returnable.
View return policy
Description
Cytochrome P450 2C18 Polyclonal antibody specifically detects Cytochrome P450 2C18 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Cytochrome P450 2C18 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| (S)-mephenytoin hydroxylase associated cytochrome P450, CPCI, CYP2C, CYP2C17, CYPIIC18, cytochrome P450 2C18, cytochrome P450, family 2, subfamily C, polypeptide 18, cytochrome P450, subfamily IIC (mephenytoin 4-hydroxylase), polypeptide 17, cytochrome P450, subfamily IIC (mephenytoin 4-hydroxylase), polypeptide 18, Cytochrome P450-6b/29c, DKFZp686I24235, EC 1.14.14.1, flavoprotein-linked monooxygenase, microsomal monooxygenase, P450-6B/29C, P450IIC17, unspecific monooxygenase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 231-490 of human CYP2C18 (NP_000763.1). NKIAENFAYIKSYVLERIKEHQESLDMNSARDFIDCFLIKMEQEKHNQQSEFTVESLIATVTDMFGAGTETTSTTLRYGLLLLLKYPEVTAKVQEEIECVVGRNRSPCMQDRSHMPYTDAVVHEIQRYIDLLPTNLPHAVTCDVKFKNYLIPKGTTIITSLTSVLHNDKEFPNPEMFDPGHFLDKSGNFKKSDYFMPFSAGKRMCMGEGLARMELFLFLTTILQNFNLKSQVDPKDIDITPIANAFGRVPPLYQLCFIPV | |
| 0.02 mL | |
| Lipid and Metabolism | |
| 1562 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction