missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome P450 2E1 Antibody (OTI5B9), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70531FR
This item is not returnable.
View return policy
Description
Cytochrome P450 2E1 Monoclonal specifically detects Cytochrome P450 2E1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Cytochrome P450 2E1 | |
| Monoclonal | |
| DyLight 680 | |
| PBS with 0.05% Sodium Azide | |
| CYP2E1 | |
| Full length recombinant protein of human Cytochrome P450 2E1 (NP_000764) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI5B9 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| 4-nitrophenol 2-hydroxylase, CPE1, CYP2Ecytochrome P450 2E1, CYPIIE1, cytochrome P450, family 2, subfamily E, polypeptide 1, cytochrome P450, subfamily IIE (ethanol-inducible), polypeptide 1, Cytochrome P450-J, EC 1.14.13.-, EC 1.14.13.n7, EC 1.14.14.1, flavoprotein-linked monooxygenase, microsomal monooxygenase, P450C2E, P450-J, xenobiotic monooxygenase | |
| Mouse | |
| Affinity Purified | |
| Lipid and Metabolism | |
| 1571.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction