missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome p450 2J2 Antibody (OTI5C9), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70533AF532
This item is not returnable.
View return policy
Description
Cytochrome p450 2J2 Monoclonal antibody specifically detects Cytochrome p450 2J2 in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| Cytochrome p450 2J2 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| Arachidonic acid epoxygenase, CPJ2, CYPIIJ2, cytochrome P450 2J2, cytochrome P450, family 2, subfamily J, polypeptide 2, cytochrome P450, subfamily IIJ (arachidonic acid epoxygenase) polypeptide 2, EC 1.14.14.1, flavoprotein-linked monooxygenase, microsomal monooxygenase | |
| Full length recombinant protein of human Cytochrome p450 2J2 (NP_000766) produced in HEK293T cells. | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 1573 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI5C9 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction