missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome P450 3A4/3A5 Antibody (F24P2B10), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50208R
This item is not returnable.
View return policy
Description
Cytochrome P450 3A4/3A5 Monoclonal specifically detects Cytochrome P450 3A4/3A5 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| Cytochrome P450 3A4, 3A5 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Ovalbumin-conjugated synthetic peptide ESRDGTVSGA | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA, Immunohistochemistry | |
| F24P2B10 | |
| Western Blot, ELISA, Immunohistochemistry | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction