missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome P450 3A4 Antibody (P6), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50207AF647
This item is not returnable.
View return policy
Description
Cytochrome P450 3A4 Monoclonal specifically detects Cytochrome P450 3A4 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications
| Cytochrome P450 3A4 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CYP3A4 | |
| Rat liver cytochrome P450 3A1 | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 1576 | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Flow Cytometry, ELISA | |
| P6 | |
| Western Blot, Flow Cytometry, ELISA | |
| Albendazole monooxygenase, Albendazole sulfoxidase, CP33, CP34, CYP3A, CYP3A3, CYPIIIA3, CYPIIIA4, Cytochrome P450 3A3, cytochrome P450 3A4, Cytochrome P450 HLp, Cytochrome P450 NF-25, cytochrome P450, family 3, subfamily A, polypeptide 4, cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 3, cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 4, Cytochrome P450-PCN1, EC 1.14.13.32, EC 1.14.13.67, EC 1.14.13.97, EC 1.14.14.1, glucocorticoid-inducible P450, HLP, MGC126680, NF-25, Nifedipine oxidase, P450C3, P450-III, steroid inducible, P450PCN1, Quinine 3-monooxygenase, Taurochenodeoxycholate 6-alpha-hydroxylase | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| P6 is highly specific for members of the CYP 3A gene family and cross reacts with CY 3A4 in human, CYP 3A in rat and potentially CYP 3A11 (based on inducibility) in mice. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction