missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome P450 3A5 Antibody (F18P3B6), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50209MFV450
This item is not returnable.
View return policy
Description
Cytochrome P450 3A5 Monoclonal antibody specifically detects Cytochrome P450 3A5 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
| Cytochrome P450 3A5 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry | |
| F18P3B6 | |
| Western Blot, ELISA, Immunohistochemistry | |
| aryl hydrocarbon hydroxylase, CP35, CYPIIIA5, cytochrome P-450, cytochrome P450 3A5, Cytochrome P450 HLp2, cytochrome P450, family 3, subfamily A, polypeptide 5, cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 5, Cytochrome P450-PCN3, DKFZp686L16231, EC 1.14.14.1, flavoprotein-linked monooxygenase, microsomal monooxygenase, niphedipine oxidase, P450PCN3FLJ31317, PCN3, xenobiotic monooxygenase | |
| Ovalbumin-conjugated synthetic peptide DSRDGTLSGE (C-terminal sequence) | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 1577 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction