missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome P450 Enzyme CYP3A1 Antibody (P6), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08767JF646
This item is not returnable.
View return policy
Description
Cytochrome P450 Enzyme CYP3A1 Monoclonal antibody specifically detects Cytochrome P450 Enzyme CYP3A1 in Human, Mouse, Rat, Porcine samples. It is validated for Western Blot, Flow Cytometry, ELISA
Specifications
| Cytochrome P450 Enzyme CYP3A1 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Human, Mouse, Rat, Pig | |
| Purified |
| Western Blot, Flow Cytometry, ELISA | |
| P6 | |
| 50mM Sodium Borate | |
| Full length protein (Rat). Rat liver cytochrome Cytochrome P450 Enzyme CYP3A1. | |
| 100 μg | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction