Learn More
Cytoglobin Mouse anti-Human, Mouse, Porcine, Rat, Clone: 4C7E11, Proteintech
Product Code. 16807255 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16807255 20 μL 20µL
16897245 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16807255 Supplier Proteintech Supplier No. 602281IG20UL
Mouse Monoclonal Antibody
Cytoglobin is a ubiquitously expressed hexacoordinate hemoglobin that may facilitate diffusion of oxygen through tissues, scavenge nitric oxide or other reactive oxygen species, or serve a protective function during oxidative stress.
Specifications
Antigen Cytoglobin
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 4C7E11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CYGB
Gene Accession No. Q8WWM9, Q921A4, Q9CX80
Gene Alias CYGB, cytoglobin, HGB, Histoglobin
Gene Symbols CYGB
Host Species Mouse
Immunogen Cytoglobin Fusion Protein Ag4161
Purification Method Protein G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100518861, 114757, 114886, 170520
Target Species Mouse, Rat, Human, Pig
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Visa mer Visa mindre
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.