missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cytohesin 1 Polyclonal Antibody
GREENER_CHOICE

Product Code. p-200128248
Change view
Click to view available options
Quantity:
20 μL
200 μL
Unit Size:
200µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17710484 200 μL 200µL
17773444 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17710484 Supplier Invitrogen™ Supplier No. PA5119069

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Cytohesin 1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP possesses an extracellular region, a single transmembrane region, and a single intracellular catalytic domains, and thus represents a receptor-type PTP. The similar gene predominately expressed in mouse brain was found to associate with, and thus regulate the activity and cellular localization of MAP kinases. The rat counterpart of this gene was reported to be regulated by the nerve growth factor, which suggested the function of this gene in neuronal growth and differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytohesin 1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene CYTH1
Gene Accession No. Q15438
Gene Alias B2-1; CLM1; CTH-1; CYTH1; CYTIP; cytoadhesin 1; cytohesin 1; CYTOHESIN-1; D17S811E; FLJ34050; FLJ41900; homolog of secretory protein SEC7; mKIAA4240; mSec7-1; PH, SEC7 and coiled-coil domain-containing protein 1; pleckstrin homology, Sec7 and coiled/coil domains 1(cytohesin 1); pleckstrin homology, Sec7 and coiled-coil domains 1; Pscd1; rSec7-1; SEC7; SEC7 homolog A; SEC7 homolog B2-1; Sec7a
Gene Symbols CYTH1
Host Species Rabbit
Immunogen Recombinant protein Cytohesin 1. The antigen corresponds to amino acid range 1-398 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100515446, 9267
Target Species Human, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.