missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 1 Antibody (3C10F7), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
€ 179.00
Specifications
| Antigen | Cytokeratin 1 |
|---|---|
| Clone | 3C10F7 |
| Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 2-5 μg/mL, Immunohistochemistry-Paraffin 1:200-1:1000, Immunohistochemistry Free-Floating |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Cytokeratin 1 Monoclonal specifically detects Cytokeratin 1 in Human, Porcine, Bovine, Canine, Equine, Feline, Goat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry Free-Floating.Specifications
| Cytokeratin 1 | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 2-5 μg/mL, Immunohistochemistry-Paraffin 1:200-1:1000, Immunohistochemistry Free-Floating | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| CK1, CK-1, cytokeratin 1, Cytokeratin-1, cytokeratin-1,67 kDa cytokeratin, EHK, EHK1, epidermolytic hyperkeratosis 1, EPPK, Hair alpha protein, K1, keratin 1, keratin, type II cytoskeletal 1, Keratin-1, KRT1A, KRTA, NEPPK, Type-II keratin Kb1 | |
| KRT1 | |
| IgG1 | |
| Unpurified | |
| CK1 (PMC01) |
| 3C10F7 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Protein Kinase | |
| Ascites with 0.03% Sodium Azide | |
| 3848 | |
| Ni-NTA purified truncated recombinant CK1 expressed in E. Coli strain BL21 (DE3) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title