missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 1 Antibody (KRT1/8342R) [Janelia Fluor™ 669], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-26957JF669
This item is not returnable.
View return policy
Description
Cytokeratin 1 Monoclonal antibody specifically detects Cytokeratin 1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Cytokeratin 1 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| CK1, CK-1, cytokeratin 1, Cytokeratin-1, cytokeratin-1,67 kDa cytokeratin, EHK, EHK1, epidermolytic hyperkeratosis 1, EPPK, Hair alpha protein, K1, keratin 1, keratin, type II cytoskeletal 1, Keratin-1, KRT1A, KRTA, NEPPK, Type-II keratin Kb1 | |
| Recombinant full-length human Cytokeratin 1 protein | |
| 0.1 mL | |
| Cell Biology, Cellular Markers, Protein Kinase, Signal Transduction | |
| 3848 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| KRT1/8342R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction