missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 1 Antibody (LHK1), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08474AF405
This item is not returnable.
View return policy
Description
Cytokeratin 1 Monoclonal antibody specifically detects Cytokeratin 1 in Human, Rat samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Cytokeratin 1 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| CK1, CK-1, cytokeratin 1, Cytokeratin-1, cytokeratin-1,67 kDa cytokeratin, EHK, EHK1, epidermolytic hyperkeratosis 1, EPPK, Hair alpha protein, K1, keratin 1, keratin, type II cytoskeletal 1, Keratin-1, KRT1A, KRTA, NEPPK, Type-II keratin Kb1 | |
| A 13-amino acid peptide (VRFVSTSYSGVTR) from the C-terminus of human Cytokeratin 1 protein (Uniprot: P04264) | |
| 100 μg | |
| Cell Biology, Cellular Markers, Protein Kinase, Signal Transduction | |
| 3848 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Immunohistochemistry (Paraffin) | |
| LHK1 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction