missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 10/13 Antibody (DE-K13), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11497FR
This item is not returnable.
View return policy
Description
Cytokeratin 10/13 Monoclonal antibody specifically detects Cytokeratin 10/13 in Human, Feline samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Cytokeratin 10/13 | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Feline | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DE-K13 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| BCIE, BIE, CK10, CK-10, cytokeratin 10, Cytokeratin-10, EHK, K10keratosis palmaris et plantaris, keratin 10, keratin, type I cytoskeletal 10, keratin-10, KPP | |
| Cytoskeletal preparation extracted from human ectocervical epithelium | |
| 0.1 mL | |
| Cell Biology, Cellular Markers | |
| 3858 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction