missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 10 Ab-2 Mouse Monoclonal Antibody, Epredia™
Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunofluorescence experiments.
€ 110.00 - € 575.00
Specifications
Antigen | Cytokeratin 10 Ab-2 |
---|---|
Clone | DE-K10 |
Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Host Species | Mouse |
Product Code | Brand | Form | Conjugate | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Conjugate | Quantity | Price | Quantity & Availability | |||
12681757
|
Epredia
MS-611-B1 |
Concentrated | Biotin | 500 μL |
€ 378.00
500µL |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12651757
|
Epredia
MS-611-B |
Concentrated | Biotin | 1 mL |
€ 575.00
1mL |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12611767
|
Epredia
MS-611-P1ABX |
Concentrated | Unconjugated | 100 μL |
€ 308.00
100µL |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12601767
|
Epredia
MS-611-P1 |
Concentrated | Unconjugated | 500 μL |
€ 272.00
500µL |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12661757
|
Epredia
MS-611-B0 |
Concentrated | Biotin | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12621767
|
Epredia
MS-611-PABX |
Concentrated | Unconjugated | 200 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12691757
|
Epredia
MS-611-P0 |
Concentrated | Unconjugated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12671757
|
Epredia
MS-611-P |
Concentrated | Unconjugated | 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12661767
|
Epredia
MS-611-R7 |
Prediluted | Unconjugated | 7 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
CK10 (Cytokeratin 10) is expressed in all suprabasal layers of the epidermis. In the epidermis, expression of CK10 strictly parallels the extent of differentiation; it is absent in the basal layer, appears in the first suprabasal layers and increases in concentration towards the granular layer. Expression of CK10 is related to maturation of malignant keratinocytes, being preferentially detected in more-differentiated parts.Host Species: Mouse
Clone: DE-K10
Isotype: IgG1
Species Reactivity: Human, Rat, Dog and Cat. Others not known.
Epitope: Not determined
Immunogen: Cytoskeletal preparation extracted from human ectocervical epithelium
Molecular Weight: 56.5 kDa
Positive Control: A431 cells, Skin
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
Specifications
Cytokeratin 10 Ab-2 | |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Mouse | |
Cytoskeletal Protein | |
Cytoskeletal preparation extracted from human ectocervical epithelium | |
Primary |
DE-K10 | |
Monoclonal | |
IVD | |
Canine, Feline, Human, Rat | |
IgG1 |