missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin 10 Antibody (KRT10/844), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-47825FR

Product Code. 18659179

  • € 588.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Cytokeratin 10 Monoclonal antibody specifically detects Cytokeratin 10 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Flow (Intracellular), CyTOF
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cytokeratin 10
Monoclonal
DyLight 680
BCIE, BIE, CK10, CK-10, cytokeratin 10, Cytokeratin-10, EHK, K10keratosis palmaris et plantaris, keratin 10, Keratin 10 (epidermolytic hyperkeratosis, keratosis palmaris et plantaris), keratin, type I cytoskeletal 10, keratin-10, KPP
Recombinant full-length human Cytokeratin 10 protein (Uniprot: P13645)
0.1 mL
Cell Biology, Cellular Markers
3858
Store at 4°C in the dark.
IgG1 κ
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Flow Cytometry (Intracellular Staining)
KRT10/844
50 mM Sodium Borate
Mouse
Protein A or G purified
RUO
Primary
Human, Mouse
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.