missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 10 Antibody (rKRT10/6923) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-20721JF669
This item is not returnable.
View return policy
Description
Cytokeratin 10 Monoclonal antibody specifically detects Cytokeratin 10 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Cytokeratin 10 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| BCIE, BIE, CK10, CK-10, cytokeratin 10, Cytokeratin-10, EHK, K10keratosis palmaris et plantaris, keratin 10, Keratin 10 (epidermolytic hyperkeratosis; keratosis palmaris et plantaris), keratin, type I cytoskeletal 10, keratin-10, KPP | |
| Recombinant fragment (around aa384-584) of human Cytokeratin 10 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Cell Biology, Cellular Markers | |
| 3858 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| rKRT10/6923 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction