missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 10 Antibody (SPM623), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54332C
This item is not returnable.
View return policy
Description
Cytokeratin 10 Monoclonal antibody specifically detects Cytokeratin 10 in Human, Canine, Feline samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF
Specifications
| Cytokeratin 10 | |
| Monoclonal | |
| DyLight 650 | |
| BCIE, BIE, CK10, CK-10, cytokeratin 10, Cytokeratin-10, EHK, K10keratosis palmaris et plantaris, keratin 10, Keratin 10 (epidermolytic hyperkeratosis, keratosis palmaris et plantaris), keratin, type I cytoskeletal 10, keratin-10, KPP | |
| Cytoskeletal preparation extracted from human ectocervical epithelium (Uniprot: P13645) | |
| 100 μg | |
| Cell Biology, Cellular Markers | |
| 3858 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
| SPM623 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Canine, Feline | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction