missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 10 Antibody (VIK-10), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-354AF405
This item is not returnable.
View return policy
Description
Cytokeratin 10 Monoclonal specifically detects Cytokeratin 10 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Cytokeratin 10 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| BCIE, BIE, CK10, CK-10, cytokeratin 10, Cytokeratin-10, EHK, K10keratosis palmaris et plantaris, keratin 10, Keratin 10 (epidermolytic hyperkeratosis; keratosis palmaris et plantaris), keratin, type I cytoskeletal 10, keratin-10, KPP | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The antibody VIK-10 reacts with Cytokeratin 10 (56.5 kDa). Cytokeratins are a member of intermediate filaments subfamily represented in epithelial tissues. | |
| Store at 4C in the dark. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| VIK-10 | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| KRT10 | |
| Cytoskeleton preparation extracted from human epidermis by detergent/high salt extraction. | |
| 0.1 mL | |
| Cytoskeleton Markers | |
| 3858 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction