Learn More
Cytokeratin 12 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-33197-100ul
 View more versions of this product
Product Code. 30232443
  • ONLINE EXCLUSIVE PRICE
    € 541.00
    € 511.35 / 100µL
    Save € 29.65
    5% Off
Description
Cytokeratin 12 Monoclonal antibody specifically detects Cytokeratin 12 in Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cytokeratin 12
Monoclonal
Western Blot 1:100 - 1:500, ELISA Recommended starting concentration is 1 μg/mL, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
keratin, type I cytoskeletal 12, CK12, K12, keratin 12
Recombinant fusion protein containing a sequence corresponding to amino acids 250-494 of human Cytokeratin 12 (KRT12) (NP_000214.1).,, Sequence:, EMQIESLNEELAYMKKNHEDELQSFRVGGPGEVSVEMDAAPGVDLTRLLNDMRAQYETIAEQNRKDAEAWFIEKSGELRKEISTNTEQLQSSKSEVTDLRRAFQNLEIELQSQLAMKKSLEDSLAEAEGDYCAQLSQVQQLISNLEAQLLQVRADAERQNVDHQRLLNVKARLELEIETYRRLLDGEAQGDGLEESLFVTDSKSQAQSTDSSKDPTKTRKIKTVVQEMVNGEVVSSQVQEIEELM
100 μL
Cell Biology, Cytoskeleton Markers, Neuroscience, Signal Transduction
3859
Store at -20°C. Avoid freeze-thaw cycles.
IgG
ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
Unconjugated
PBS (pH 7.3), 50% glycerol, 0.05% BSA
Rabbit
Affinity purified
RUO
Primary
Mouse, Rat