missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 13 Antibody (KRT13/2659), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08284V
This item is not returnable.
View return policy
Description
Cytokeratin 13 Monoclonal antibody specifically detects Cytokeratin 13 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Cytokeratin 13 | |
| Monoclonal | |
| DyLight 405 | |
| CK13, CK-13, cytokeratin 13, Cytokeratin-13, K13cytokeratin-13, keratin 13, keratin, type I cytoskeletal 13, keratin-13, MGC161462, MGC3781 | |
| Recombinant full-length human Cytokeratin 13 protein (Uniprot: P13646) | |
| 100 μg | |
| Cell Biology, Cellular Markers | |
| 3860 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| KRT13/2659 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction