missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 13 Antibody (KRT13/2850), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08288JF646
This item is not returnable.
View return policy
Description
Cytokeratin 13 Monoclonal antibody specifically detects Cytokeratin 13 in Human, Mouse, Rat, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Cytokeratin 13 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| CK13, CK-13, cytokeratin 13, Cytokeratin-13, K13cytokeratin-13, keratin 13, keratin, type I cytoskeletal 13, keratin-13, MGC161462, MGC3781 | |
| esophageal keratins of rabbit origin | |
| 100 μg | |
| Cell Biology, Cellular Markers | |
| 3860 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| KRT13/2850 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Rabbit | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction