missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cytokeratin 13 Recombinant Rabbit Monoclonal Antibody (5M6B10)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA537816
This item is not returnable.
View return policy
Description
Positive Samples: Mouse lung, Mouse uterus Immunogen sequence: LTGNEKITMQ NLNDRLASYL EKVRALEEAN ADLEVKIRDW HLKQSPASPE RDYSPYYKTI EELRDKILTA TIENNRVILE IDNARLAADD FRLKYENELA L.
Cytokeratins are a subfamily of intermediate filament proteins and are characterized by a remarkable biochemical diversity, represented in human epithelial tissues by at least 20 different polypeptides. They range in molecular weight between ~40 kDa and ~68 kDa and isoelectric pH between 4.9 - 7.8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial malignancies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays.
Specifications
| Cytokeratin 13 | |
| Recombinant Monoclonal | |
| 0.1 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P08730, P13646, Q6IFV4 | |
| KRT13 | |
| Synthetic peptide. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 5M6B10 | |
| Unconjugated | |
| KRT13 | |
| 47 kDa cytokeratin; CK13; CK-13; cytokeratin 13; cytokeratin-13; K13; K-13; Ka13; keratin 13; keratin 13, type I; keratin complex 1, acidic, gene 13; keratin, type I cytoskeletal 13; Keratin-13; Krt-1.13; Krt1-13; KRT13; krt13 {ECO:0000250; MGC161462; MGC3781; Type I keratin Ka13; UniProtKB:P13646}; WSN2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 16663, 287699, 3860 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction