missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 14 Antibody (KRT14/6987R), PerCP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20731PCP
This item is not returnable.
View return policy
Description
Cytokeratin 14 Monoclonal antibody specifically detects Cytokeratin 14 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Cytokeratin 14 | |
| Monoclonal | |
| PerCP | |
| CK14, CK-14, cytokeratin 14, cytokeratin-14, EBS3, EBS4, K14, keratin 14, keratin 14 (epidermolysis bullosa simplex, Dowling-Meara, Koebner), keratin, type I cytoskeletal 14, Keratin-14, NFJ | |
| Recombinant human Cytokeratin 14 fragment (around aa350-472) (exact sequence is proprietary) | |
| 0.1 mL | |
| Cell Biology, Cellular Markers | |
| 3861 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| KRT14/6987R | |
| PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction