missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 15 Antibody (KRT15/2103R), Alexa Fluor™ 532, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08609AF532
This item is not returnable.
View return policy
Description
Cytokeratin 15 Monoclonal antibody specifically detects Cytokeratin 15 in Human, Rat, Bovine samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin)
Specifications
| Cytokeratin 15 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| CK15, CK-15, cytokeratin 15, cytokeratin-15, K15keratin-15, basic, K1CO, keratin 15, keratin, type I cytoskeletal 15, Keratin-15, keratin-15, beta, KRTB, type I cytoskeletal 15 | |
| Recombinant human full-length Cytokeratin 15 protein (Uniprot: P19012) | |
| 100 μg | |
| Cell Biology, Cellular Markers | |
| 3866 | |
| Store at 4°C in the dark. | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Paraffin) | |
| KRT15/2103R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Rat, Bovine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction