missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 15 Antibody (SPM190), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54453AF594
This item is not returnable.
View return policy
Description
Cytokeratin 15 Monoclonal antibody specifically detects Cytokeratin 15 in Human, Rat, Bovine samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), CyTOF
Specifications
| Cytokeratin 15 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| CK15, CK-15, cytokeratin 15, cytokeratin-15, K15keratin-15, basic, K1CO, keratin 15, keratin, type I cytoskeletal 15, Keratin-15, keratin-15, beta, KRTB, type I cytoskeletal 15 | |
| A 17 amino acids synthetic peptide from the C- terminal of Human Cytokeratin 15 (Uniprot: P19012) | |
| 0.1 mL | |
| Cell Biology, Cellular Markers | |
| 3866 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), CyTOF | |
| SPM190 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Rat, Bovine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction