missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 16 Antibody (LL025), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08684APC
This item is not returnable.
View return policy
Description
Cytokeratin 16 Monoclonal antibody specifically detects Cytokeratin 16 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Cytokeratin 16 | |
| Monoclonal | |
| APC | |
| CK16, CK-16, cytokeratin 16, cytokeratin-16, FNEPPK, focal non-epidermolytic palmoplantar keratoderma, K16, K1CP, keratin 16, keratin, type I cytoskeletal 16, keratin-16, KRT16A, NEPPK | |
| A synthetic peptide from the C-terminal of human Cytokeratin 16 (Uniprot: P08779) | |
| 0.1 mL | |
| Cell Biology, Cellular Markers | |
| 3868 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| LL025 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction