missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 17 Antibody (KRT17/4604), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08541UV
This item is not returnable.
View return policy
Description
Cytokeratin 17 Monoclonal antibody specifically detects Cytokeratin 17 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Cytokeratin 17 | |
| Monoclonal | |
| DyLight 350 | |
| 39.1, CK-17, cytokeratin-17, K17, keratin 17, keratin, type I cytoskeletal 17, keratin-17, PC, PC2, PCHC1 | |
| Recombinant full-length human Cytokeratin 17 protein (Uniprot: Q04695) | |
| 100 μg | |
| Cancer | |
| 3872 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| KRT17/4604 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction