missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 17 Antibody (KRT17/778), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47684FR
This item is not returnable.
View return policy
Description
Cytokeratin 17 Monoclonal antibody specifically detects Cytokeratin 17 in Human, Rat, Porcine, Bovine, Goat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Cytokeratin 17 | |
| Monoclonal | |
| DyLight 680 | |
| 39.1, CK-17, cytokeratin-17, K17, keratin 17, keratin, type I cytoskeletal 17, keratin-17, PC, PC2, PCHC1 | |
| Recombinant full-length human Cytokeratin 17 protein (Uniprot: Q04695) | |
| 0.1 mL | |
| Cancer | |
| 3872 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| KRT17/778 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Rat, Pig, Bovine, Goat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction