Learn More
Cytokeratin 18 Antibody (C-04 (same as Ks18.04)), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-11485DL594
Product Code. 18529826
  • ONLINE EXCLUSIVE PRICE
    € 774.00
    € 731.85 / 0.10mL
    Save € 42.15
    5% Off
Description
Cytokeratin 18 Monoclonal antibody specifically detects Cytokeratin 18 in Human, Mouse, Rat, Porcine, Bovine, Canine, Equine, Feline, Goat, Hamster, Ferret, Monkey, Sheep, Chicken (Negative), Xenopus (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cytokeratin 18
Monoclonal
DyLight 594
50mM Sodium Borate
Mouse
Protein A or G purified
RUO
Primary
Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Feline, Goat, Hamster, Ferret, Monkey, Sheep, Chicken (Negative), Xenopus (Negative)
Purified
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
C-04 (same as Ks18.04)
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Cell proliferation-inducing gene 46 protein, cell proliferation-inducing protein 46, CK-18, CYK18, cytokeratin 18, cytokeratin-18, K18, keratin 18, keratin, type I cytoskeletal 18, keratin-18
Cytoskeletal preparation extracted from the human vulvar squamous carcinoma cell line A431
0.1 mL
Cancer, Cell Biology, Cellular Markers, Phospho Specific
3875
Store at 4°C in the dark.
IgG1 κ