missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 18 Antibody (KRT18/835), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47986AF405
This item is not returnable.
View return policy
Description
Cytokeratin 18 Monoclonal antibody specifically detects Cytokeratin 18 in Human, Bovine (Negative), Canine (Negative), Hamster (Negative), Porcine (Negative), Rat (Negative), Sheep (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array
Specifications
| Cytokeratin 18 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| Cell proliferation-inducing gene 46 protein, cell proliferation-inducing protein 46, CK-18, CYK18, cytokeratin 18, cytokeratin-18, K18, keratin 18, keratin, type I cytoskeletal 18, keratin-18 | |
| Recombinant full-length human Cytokeratin 18 protein (Uniprot: P05783) | |
| 0.1 mL | |
| Cancer, Cell Biology, Cellular Markers | |
| 3875 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array | |
| KRT18/835 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Bovine (Negative), Canine (Negative), Hamster (Negative), Porcine (Negative) | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction