missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 18 Antibody (LDK18) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 214.00 - € 377.00
Specifications
| Antigen | Cytokeratin 18 |
|---|---|
| Clone | LDK18 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18625958
|
Novus Biologicals
NBP2-50463-0.025ml |
0.025 mL |
€ 214.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18601169
|
Novus Biologicals
NBP2-50463 |
0.1 mL |
€ 377.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cytokeratin 18 Monoclonal antibody specifically detects Cytokeratin 18 in Human, Mouse, Porcine samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, CyTOFSpecifications
| Cytokeratin 18 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cancer, Cell Biology, Cellular Markers | |
| PBS | |
| 3875 | |
| IgG1 | |
| Protein A purified |
| LDK18 | |
| Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Pig | |
| Cell proliferation-inducing gene 46 protein, cell proliferation-inducing protein 46, CK-18, CYK18, cytokeratin 18, cytokeratin-18, K18, keratin 18, keratin, type I cytoskeletal 18, keratin-18 | |
| Synthetic peptide of human Cytokeratin 18 (CSETNDTKVLRH-COOH) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title