missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 18 Antibody (LDK18), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50463FR
This item is not returnable.
View return policy
Description
Cytokeratin 18 Monoclonal specifically detects Cytokeratin 18 in Human, Mouse, Porcine samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry.
Specifications
| Cytokeratin 18 | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| KRT18 | |
| Synthetic peptide of human Cytokeratin 18 (CSETNDTKVLRH-COOH) | |
| 0.1 mL | |
| Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers | |
| 3875 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
| LDK18 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
| Cell proliferation-inducing gene 46 protein, cell proliferation-inducing protein 46, CK-18, CYK18, cytokeratin 18, cytokeratin-18, K18, keratin 18, keratin, type I cytoskeletal 18, keratin-18 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Pig | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction