missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 18 Antibody (OTI1E1), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71086AF750
This item is not returnable.
View return policy
Description
Cytokeratin 18 Monoclonal specifically detects Cytokeratin 18 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Cytokeratin 18 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| KRT18 | |
| Recombinant protein fragment corresponding to amino acids 69-372 of human Cytokeratin 18 (NP_000215) produced in E.coli. | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI1E1 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Cell proliferation-inducing gene 46 protein, cell proliferation-inducing protein 46, CK-18, CYK18, cytokeratin 18, cytokeratin-18, K18, keratin 18, keratin, type I cytoskeletal 18, keratin-18 | |
| Mouse | |
| 0.1 mL | |
| 3875 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction