missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 18 Antibody (OTI1E1), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71086APC
This item is not returnable.
View return policy
Description
Cytokeratin 18 Monoclonal antibody specifically detects Cytokeratin 18 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| Cytokeratin 18 | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry | |
| OTI1E1 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
| Cell proliferation-inducing gene 46 protein, cell proliferation-inducing protein 46, CK-18, CYK18, cytokeratin 18, cytokeratin-18, K18, keratin 18, keratin, type I cytoskeletal 18, keratin-18 | |
| Human recombinant protein fragment corresponding to amino acids 69-372 of human CK18 (NP_000215) produced in E.coli. | |
| 0.1 mL | |
| Cancer, Cell Biology, Cellular Markers, Phospho Specific | |
| 3875 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction