missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 18 Antibody (RGE53), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-97715AF700
This item is not returnable.
View return policy
Description
Cytokeratin 18 Monoclonal specifically detects Cytokeratin 18 in Human, Mouse, Rat, Porcine, Canine, Chicken, Guinea Pig, Hamster, Rabbit, Zebrafish samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
Specifications
| Cytokeratin 18 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| Cell proliferation-inducing gene 46 protein, cell proliferation-inducing protein 46, CK-18, CYK18, cytokeratin 18, cytokeratin-18, K18, keratin 18, keratin, type I cytoskeletal 18, keratin-18 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| RGE53 reacts exclusively with cytokeratin 18 which is present in glandular epithelial cells of the digestive, respiratory, and urogenital tracts, endocrine and exocrine cells and mesothelial cells, as well as adenocarcinomas originating from them. | |
| Store at 4C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| RGE53 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
| KRT18 | |
| RGE53 is a mouse monoclonal IgG1 antibody derived by fusion of SP2/0-Ag14 mouse myeloma cells with spleen cells from a BALB/c mouse immunized with a cytoskeletal preparation of HeLa cells. | |
| 0.1 mL | |
| Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Phospho Specific | |
| 3875 | |
| Human, Mouse, Rat, Pig, Canine, Chicken, Guinea Pig, Hamster, Rabbit, Zebrafish | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction