missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 18 Antibody (rKRT18/7299), Novus Biologicals™
Mouse Monoclonal Antibody
€ 249.00 - € 494.00
Specifications
| Antigen | Cytokeratin 18 |
|---|---|
| Clone | rKRT18/7299 |
| Concentration | 0.2 mg/ml |
| Dilution | Western Blot 1-2 μg/mL, Flow Cytometry 1-2 μg/million cells, Immunocytochemistry/Immunofluorescence 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18670295
|
Novus Biologicals
NBP3-20333-20ug |
20 μg |
€ 249.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18679514
|
Novus Biologicals
NBP3-20333-100ug |
100 μg |
€ 494.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cytokeratin 18 Monoclonal antibody specifically detects Cytokeratin 18 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| Cytokeratin 18 | |
| 0.2 mg/ml | |
| Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Cell Biology, Cellular Markers, Phospho Specific | |
| 10mM PBS with 0.05% BSA | |
| 3875 | |
| IgG1 κ | |
| Protein A or G purified |
| rKRT18/7299 | |
| Western Blot 1-2 μg/mL, Flow Cytometry 1-2 μg/million cells, Immunocytochemistry/Immunofluorescence 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Cell proliferation-inducing gene 46 protein, cell proliferation-inducing protein 46, CK-18, CYK18, cytokeratin 18, cytokeratin-18, K18, keratin 18, keratin, type I cytoskeletal 18, keratin-18 | |
| Recombinant full-length human Cytokeratin 18 protein | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title