missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 18 Mouse, Clone: DE-K18, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 3875-MSM3-P1
This item is not returnable.
View return policy
Specifications
| Cytokeratin 18 | |
| Cytoplasmic | |
| DE-K18 | |
| Unconjugated | |
| NP_000215 | |
| Mouse | |
| 45kDa | |
| Protein A/G Purified | |
| RUO | |
| 3875 | |
| IgG1 κ |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide | |
| KRT18 | |
| Cytoskeletal preparation extracted from the human vulvar squamous carcinoma cell line A431 | |
| MCF-7, HeLa cells, Breast Cancer | |
| 100 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction